Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLEC10 Monoclonal Antibody (4F9D7)
GREENER_CHOICE

Catalog No. FERMA549031
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549031 Supplier Invitrogen™ Supplier No. MA549031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COLEC10 Monoclonal Antibody for Inhibition, Western Blot

COLEC10 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLEC10
Applications Inhibition Assays, Western Blot
Classification Monoclonal
Clone 4F9D7
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene COLEC10
Gene Accession No. Q9Y6Z7
Gene Alias CL-34; CLL1; CL-L1; Colec10; collectin liver 1; collectin liver protein 1; collectin subfamily member 10; collectin sub-family member 10; collectin sub-family member 10 (C-type lectin); collectin-10; Collectin-34; collectin-L1; UNQ366/PRO702
Gene Symbols COLEC10
Host Species Mouse
Immunogen Antibody 4F9D7 recognizes human Collectin-10 (CL-L1).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10584
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.