Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLEC10 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5122077
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122077 100 μL
PIPA5122078 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122077 Supplier Invitrogen™ Supplier No. PA5122077
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COLEC10 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLEC10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene COLEC10
Gene Accession No. Q9Y6Z7
Gene Alias CL-34; CLL1; CL-L1; Colec10; collectin liver 1; collectin liver protein 1; collectin subfamily member 10; collectin sub-family member 10; collectin sub-family member 10 (C-type lectin); collectin-10; Collectin-34; collectin-L1; UNQ366/PRO702
Gene Symbols COLEC10
Host Species Rabbit
Immunogen Pro34-Lys277 (Accession Q9Y6Z7), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10584
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.