Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLEC11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546802 Supplier Invitrogen™ Supplier No. PA546802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AQAGLARVFI GINDLEKEGA FVYSDHSPMR TFNKWRSGEP NNAYDEEDCV.

This gene encodes a member of the collectin family of C-type lectins that possess collagen-like sequences and carbohydrate recognition domains. Collectins are secreted proteins that play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. The encoded protein binds to multiple sugars with a preference for fucose and mannose. Mutations in this gene are a cause of 3MC syndrome-2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLEC11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene COLEC11
Gene Accession No. Q9BWP8
Gene Alias 1010001H16Rik; 3MC2; CLK1; CL-K1; CL-K1-I; CL-K1-II; CL-K1-IIa; CL-K1-IIb; Colec11; Collectin 11; Collectin K1; collectin kidney protein 1; collectin subfamily member 11; collectin sub-family member 11; Collectin11; collectin-11; hypothetical protein LOC492459; UNQ596/PRO1182; wu:fb82e09; zgc:103572
Gene Symbols COLEC11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human COLEC11 (aa 194-243).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78989
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.