Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLEC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111263 Supplier Invitrogen™ Supplier No. PA5111263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITTISQANEQ NLKDLQDLHK DAENRTAIKF NQLEERFQLF ETDIVNIISN ISYTAHHLRT LTSNLNEVRT TCTDTLTKHT DDLTSLNNTL ANIRLDSVSL RMQQDLMRSR LDTEVANLSV IMEEMKLVDS KHGQL Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 90%.

This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor, a cell surface glycoprotein that can bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. In addition, these receptors can recognize oxidized phospholipids so they may also participate in removing oxidatively damaged or apoptotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLEC12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Colec12
Gene Accession No. Q5KU26
Gene Alias CLP1; CL-P1; COLEC12; Collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; Scara4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; Srcl
Gene Symbols Colec12
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human COLEC12. Recombinant protein control fragment (Product #RP-90348).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.