Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLGALT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556845 Supplier Invitrogen™ Supplier No. PA556845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFSSRQAGIQ MYLCNREHYG YLPIPLKPHQ TLQEDIENLI HVQIEAMIDR PPMEPSQYVS VVPKYPDKMG FDEI Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

GLT25D2 has a beta-galactosyltransferase activity and transfers beta-galactose to hydroxylysine residues on collagen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLGALT2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COLGALT2
Gene Accession No. Q6NVG7, Q8IYK4
Gene Alias AI427652; C1orf17; ColGalT 2; Colgalt2; Collagen beta(1-O)galactosyltransferase 2; D730024P12; Glt25d2; glycosyltransferase 25 domain containing 2; glycosyltransferase 25 family member 2; Hydroxylysine galactosyltransferase 2; KIAA0584; Procollagen galactosyltransferase; procollagen galactosyltransferase 2; RGD1304702
Gene Symbols COLGALT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COLGALT2. Recombinant protein control fragment (Product #RP-94566).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23127, 269132, 289081
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.