Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COLGALT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556846
Description
Immunogen sequence: PTHYTGQPGY LSDTETSTIW DNETVATDWD RTHAWKSRKQ SRIYSNAKNT EALPPPTSLD TVPSRDEL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
GLT25D2 has a beta-galactosyltransferase activity and transfers beta-galactose to hydroxylysine residues on collagen.
Specifications
COLGALT2 | |
Polyclonal | |
Unconjugated | |
COLGALT2 | |
AI427652; C1orf17; ColGalT 2; Colgalt2; Collagen beta(1-O)galactosyltransferase 2; D730024P12; Glt25d2; glycosyltransferase 25 domain containing 2; glycosyltransferase 25 family member 2; Hydroxylysine galactosyltransferase 2; KIAA0584; Procollagen galactosyltransferase; procollagen galactosyltransferase 2; RGD1304702 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23127, 269132, 289081 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NVG7, Q8IYK4 | |
COLGALT2 | |
Recombinant protein corresponding to Human COLGALT2. Recombinant protein control fragment (Product #RP-94565). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction