Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLGALT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109571 Supplier Invitrogen™ Supplier No. PA5109571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RFVAERDSED DGEEPVVFPE SPLQSPTVLV AVLARNAAHT LPHFLGCLER LDYPKSRMAI WAATDHNVDN TTEIFREWLK NVQRLYHYVE WRPMDEPESY PDEIGPKHWP TSRFAHVMKL RQAALRTARE KWSDYILFID VDNFLTNPQT LNLLIAENKT IVAPMLESRG LYSNFWCGIT PKGFYKRTPD YVQIREWKRT GCF.

GLT25D2 has a beta-galactosyltransferase activity and transfers beta-galactose to hydroxylysine residues on collagen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLGALT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene COLGALT2
Gene Accession No. Q6NVG7, Q8IYK4
Gene Alias AI427652; C1orf17; ColGalT 2; Colgalt2; Collagen beta(1-O)galactosyltransferase 2; D730024P12; Glt25d2; glycosyltransferase 25 domain containing 2; glycosyltransferase 25 family member 2; Hydroxylysine galactosyltransferase 2; KIAA0584; Procollagen galactosyltransferase; procollagen galactosyltransferase 2; RGD1304702
Gene Symbols COLGALT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-230 of human COLGALT2 (NP_055916.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23127, 269132
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.