Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COLGALT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546014 Supplier Invitrogen™ Supplier No. PA546014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YTGQPGYLSD TETSTIWDNE TVATDWDRTH AWKSRKQSRI YSNAKNTEAL Sequence homology: Cow: 86%; Dog: 79%; Horse: 86%; Human: 100%; Pig: 79%; Rat: 79%.

GLT25D2 has a beta-galactosyltransferase activity and transfers beta-galactose to hydroxylysine residues on collagen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLGALT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene COLGALT2
Gene Accession No. Q8IYK4
Gene Alias AI427652; C1orf17; ColGalT 2; Colgalt2; Collagen beta(1-O)galactosyltransferase 2; D730024P12; Glt25d2; glycosyltransferase 25 domain containing 2; glycosyltransferase 25 family member 2; Hydroxylysine galactosyltransferase 2; KIAA0584; Procollagen galactosyltransferase; procollagen galactosyltransferase 2; RGD1304702
Gene Symbols COLGALT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GLT25D2 (aa 562-611).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23127
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.