Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen 6 alpha 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980350
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0350 Supplier Bioss Supplier No. BS0553R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The extensive family of COL gene products(collagens) iscomposed ofseveral chain types, including fibril-forming interstitialcollagens(typesI, II, III and V) and basement membrane collagens(type IV), each type containing multiple isoforms. Collagens are fibrous, extracellularmatrix proteinswith high tensile strength and are themajorcomponents ofconnective tissue,such astendons and cartilage. Allcollagenscontain a triple helix domain and frequentlyshow lateralself-association in order to formcomplexconnective tissues. Several collagens also play a role in cell adhesion, important formaintaining normal tissue architecture and function.

Specifications

Antigen Collagen 6 alpha 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Col6a1
Gene Accession No. P12109
Gene Alias AI747156; alpha 1 (VI) chain (61 AA); BTHLM1; Col6a1; Col6a-1; Collagen alpha-1(VI) chain; collagen type VI alpha 1; collagen VI, alpha-1 polypeptide; collagen, type VI, alpha 1; OPLL; procollagen, type VI, alpha 1; RGD1565398; UCHMD1
Gene Symbols Col6a1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Collagen VI.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1291
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.