Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen I alpha 1 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP13005402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP13005402 0.02 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP13005402 Supplier Novus Biologicals Supplier No. NBP1300540.02ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 36 publications

Collagen I alpha 1 Polyclonal specifically detects Collagen I alpha 1 in Human, Mouse, Rat, Amphibian, Avian, Mammal, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Collagen I alpha 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with No Preservative
Gene Accession No. P02452
Gene Alias Alpha-1 type I collagen, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, Collagen1, Collagen-1, OI4, pro-alpha-1 collagen type 1
Gene Symbols COL1A1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues specific to the collagen 1, alpha 1 telopeptide conjugated to KLH
Molecular Weight of Antigen 140 kDa
Purification Method Affinity Purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1277
Test Specificity Specific for the ∼ 140 kDa telopeptide portion of the collagen I alpha 1 polypeptide. The antibody works well for immunohistochemistry on paraformaldehyde-fixed sections with a simple antigen-retrieval protocol (incubate slides for 20 minutes at 90 degrees C in 10 mM sodium citrate (pH 6.0)/ 0.1 % Tween-20). Note that in paraffin sections of formaldehydefixed fibrotic mouse lung tissue, the antibody recognizes mature collagen I that has formed fibrils in the extracellular matrix.
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.