Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen I alpha 1 Antibody (COL-1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 35 publications
Supplier: Novus Biologicals NB6004500.02ML
Description
Collagen I alpha 1 Monoclonal specifically detects Collagen I alpha 1 in Human, Mouse, Rat, Porcine, Bovine, Equine, Feline, Mammal, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Specifications
Collagen I alpha 1 | |
Monoclonal | |
Unconjugated | |
Ascites with 15mM Sodium Azide | |
COL1A1 | |
Full length bovine native protein (purified). | |
Unpurified | |
RUO | |
Primary | |
Recognizes the native (helical) form of collagen type I using ELISA and dot-blot. Does not react with the thermally-denatured molecule. The shows no cross-reactivity with collagen types II, III, IV, V, VI, VII, IX, X and XI. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
COL-1 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:2000, Dot Blot 1:100-1:2000, Immunocytochemistry | |
Alpha-1 type I collagen, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, Collagen1, Collagen-1, OI4, pro-alpha-1 collagen type 1 | |
Mouse | |
139 kDa | |
0.02 mL | |
Cytoskeleton Markers, Extracellular Matrix | |
1277 | |
Human, Mouse, Rat, Pig, Bovine, Equine, Mammalia, Rabbit | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction