Learn More
Collagen I Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 195 publications
Supplier:  Novus Biologicals NB6004080.025MG
Catalog No. NB600408025
Only null left
Add to cart
Description
Description
Collagen I Polyclonal specifically detects Collagen I in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, SDS-Page, Fluorophore-linked immunosorbent assay, Microarray.
Specifications
Specifications
Collagen I alpha 1
Polyclonal
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Alpha-1 type I collagen, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, Collagen1, Collagen-1, OI4, pro-alpha-1 collagen type 1
Rabbit
139 kDa
0.025 mg
Cancer, Cell Cycle and Replication, Cytoskeleton Markers, Extracellular Matrix
1277
Human, Mouse, Rat, Pig, Bovine, Feline, Rabbit
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1:1000, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry 1:1600
P02452, P02453
COL1A1
Collagen Type I from human and bovine placenta by limited pepsin digestion and selective salt precipitation.
Affinity Purified
RUO
Primary
Typically less than 1% cross-reactivity against other types of collagens was detected by ELISA against purified standards. Some class-specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type I collagens and has negligible cross-reactivity with Type II, III, IV, V or VI collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only