Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen I/II/III Recombinant Rabbit Monoclonal Antibody (HL2048/HL1907), Invitrogen™

Catalog No. FEMA550521
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550521 Supplier Thermo Scientific Supplier No. MA550521

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Recombinant Monoclonal Antibody

Collagen I/II/III is a gene target that codes for various types of collagen proteins. Collagen is the most abundant protein in the human body and is found in bones, skin, tendons, cartilage, and other connective tissues. Collagen I is the most common type of collagen and is responsible for providing strength and structure to bones, tendons, and skin. Collagen II is found in cartilage and is essential for maintaining its strength and elasticity. Collagen III is also found in skin and other connective tissues and plays a role in wound healing. The gene target Collagen I/II/III is important for understanding the function and regulation of collagen production, as well as for developing therapies for conditions related to collagen deficiencies or disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen I/II/III
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2048/HL1907
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene COL1A2
Gene Accession No. P02452, P02454, P02458, P02461, P02466, P05539, P08121, P08123, P11087, P13941, P28481, Q01149
Gene Alias COL1A1/COL2A1/COL3A1
Gene Symbols COL1A1, COL1A2, COL2A1, COL3A1
Host Species Rabbit
Immunogen Recombinant fragment of human Collagen I+Collagen II+Collagen III.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277, 1278, 1280, 1281, 12824, 12825, 12842, 12843, 25412, 29393, 84032, 84352
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.