Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen I/III Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10064523C
Description
Collagen I/III Polyclonal antibody specifically detects Collagen I/III in Bovine samples. It is validated for ELISA, ImmunofluorescenceSpecifications
Collagen I/III | |
Polyclonal | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Alpha-1 type I collagen, Collagen 1/3, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, OI4, pro-alpha-1 collagen type 1 | |
Bovine collagen type I and III from skin | |
0.25 mL | |
Cell Biology, Cellular Markers, Extracellular Matrix | |
1277 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction