Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen I/III Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10064523
Description
Collagen I/III Polyclonal specifically detects Collagen I/III in Bovine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Collagen I/III | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1.0% Mannitol, 1.0% Dextran with No Preservative | |
| Alpha-1 type I collagen, Collagen 1/3, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, OI4, pro-alpha-1 collagen type 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB100-64523 reacts with both Collagen I and III. Less than 0.5% reactivity was observed with bovine albumin and immunoglobulins. Collagen is located in the extracellular matrix of connective tissues. It is part of the interacting network of proteoglycans and proteins that provides a structural framework for both soft and calcified connective tissues. | |
| Bovine | |
| IgG |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:40 | |
| P02453 | |
| COL1A1 | |
| Bovine collagen type I and III from skin | |
| 0.25 mL | |
| Cytoskeleton Markers, Extracellular Matrix | |
| 1277 | |
| Reconstitute to desired concentration. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction