Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Collagen III Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586973
Description
Purity is > 95% by SDS-PAGE.
Collagen III is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen. Mutations in th gene are associated with Ehlers Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications
Collagen I/III | |
Polyclonal | |
Unconjugated | |
COL1A2 | |
AA960264; AI325291; alpha 2 type I procollagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 (III) collagen; alpha1 chain of type I collagen; alpha-1 type 1 collagen; alpha-1 type I collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; Alpha-2 type I collagen; alpha2(I) collagen; AW550625; Col1a1; Col1a-1; COL1A1; matrix protein; COL1A2; Col1a-2; COL1A2 gene exons 8 through 12; COL3A1; Col3a-1; Cola1; Cola-1; Cola2; Cola-2; Collagen 1; collagen 1 alpha 2 chain; Collagen 3; collagen alpha 1 chain type I; collagen alpha 1 type III; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen alpha-1(III) chain; Collagen alpha-2(I) chain; collagen COL1A2; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen type I alpha 1 chain; collagen type I alpha 2 chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type I, alpha 1; collagen, type I, alpha 2; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; EDS4A; EDSC; Ehlers-Danlos syndrome type IV, autosomal dominant; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Mov13; Mov-13; Ms10w; OI1; OI2; OI3; OI4; oim; osteogenesis imperfecta; PIIINP; pro-alpha 2(I); pro-alpha-1 collagen type 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; procollagen, type III, alpha 1; similar to pro alpha 1(I) collagen; type I collagen alpha 1 chain; type I collagen alpha1; type I collagen alpha-2 chain; type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I procollagen pro-alpha 2 chain; type III collagen | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1277, 1278, 1281, 12825, 12842, 12843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P02452, P02461, P08121, P08123, P11087, Q01149 | |
COL1A1, COL1A2, Col3a1 | |
Recombinant fusion protein of human III alpha 1/COL3A1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction