Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen I/III Polyclonal Antibody, Invitrogen™

Catalog No. PIPA185328
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA185328 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA185328 Supplier Thermo Scientific Supplier No. PA185328

Rabbit Polyclonal Antibody

This antibody shows <0.5% cross-reactivity with porcine albumin and immunoglobulins. Reconstitute with 0.5 mL of distilled water or saline. Prior to reconstitution, store at 4oC. After reconstitution, store undiluted at -20oC, avoiding freeze/thaw cycles. Rabbit anti Porcine collagen I/III antibody reacts with native and heat denatured porcine collagen type 1 and 3.Less than 0.5% reactivity is observed with porcine albumin and immunoglobulins.

Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen I/III
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation 0.01M sodium borate with 0.01M Na2PO4, 0.1M NaCl, 1% dextran, 1% mannitol and no preservative
Gene Col3a1
Gene Accession No. 0
Gene Alias AA960264; AI325291; alpha 2 type I procollagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 (III) collagen; alpha1 chain of type I collagen; alpha-1 type 1 collagen; alpha-1 type I collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; Alpha-2 type I collagen; alpha2(I) collagen; AW550625; Col1a1; Col1a-1; COL1A1; matrix protein; COL1A2; Col1a-2; COL1A2 gene exons 8 through 12; COL3A1; Col3a-1; Cola1; Cola-1; Cola2; Cola-2; Collagen 1; collagen 1 alpha 2 chain; Collagen 3; collagen alpha 1 chain type I; collagen alpha 1 type III; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen alpha-1(III) chain; Collagen alpha-2(I) chain; collagen COL1A2; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen type I alpha 1 chain; collagen type I alpha 2 chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type I, alpha 1; collagen, type I, alpha 2; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; EDS4A; EDSC; Ehlers-Danlos syndrome type IV, autosomal dominant; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Mov13; Mov-13; Ms10w; OI1; OI2; OI3; OI4; oim; osteogenesis imperfecta; PIIINP; pro-alpha 2(I); pro-alpha-1 collagen type 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; procollagen, type III, alpha 1; similar to pro alpha 1(I) collagen; type I collagen alpha 1 chain; type I collagen alpha1; type I collagen alpha-2 chain; type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I procollagen pro-alpha 2 chain; type III collagen
Gene Symbols COL1A1, COL1A2, Col3a1
Host Species Rabbit
Immunogen Porcine collagen, types 1 and 3 from skin.
Purification Method affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100152001, 100626716, 100738123
Target Species Pig
Content And Storage 4° C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.