Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen II Antibody (CIIC1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311227
Description
Collagen II Monoclonal specifically detects Collagen II in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.Specifications
Collagen II | |
Monoclonal | |
0.1 mg/ml | |
Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay | |
Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC | |
The monoclonal antibody CIICI reacts with the C1 epitope (triple helical position 358-364) of collagen type II. | |
50 μg | |
Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
1280 | |
Store at 4°C. Do not freeze. | |
IgG2a |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunoassay | |
CIIC1 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction