Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen II Monoclonal Antibody (2B1.5), Invitrogen™

Catalog No. PIMA512789
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512789 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512789 Supplier Thermo Scientific Supplier No. MA512789

Mouse Monoclonal Antibody

Staining of formalin/parafin tissues requires disgestion of tissue sections with pepsin at 1 mg/mL Tris-HCl, pH 2.0 for 15 min at RT or 10 min at 37C.

COL2A1 is the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen II
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B1.5
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Col2a1
Gene Accession No. P02458, P02459, P02460, P05539, P28481
Gene Alias alpha 1 type II procollagen; alpha 1 type IIA collagen; alpha 1 type IIA collagen precursor; alpha collagen type II; alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CB11; CG2A1A; CGA; Chondrocalcin; COL11A3; Col2; Col2a; col2a1; Col2a-1; col2a1.L; col2a1a; col2a1-a; col2a1-b; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen type II alpha 1 chain; collagen type IIA; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); collagen, type II, alpha 1 L homeolog; COLLII; cyanogen bromide; Del1; disproportionate micromelia; Dmm; Lpk; M100856; MGC131516; pro alpha1 type II collagen; pro-alpha 1(II); pro-alpha-1 type II collagen; procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; Rgsc856; SEDC; STL1; type II collagen; type II collagen cyanogen bromide fragment CB10; type II collogen cyanogen bromide fragment CB8; unnamed protein product; X-col II; xcol2a1; XELAEV_18013523mg
Gene Symbols Col2a1
Host Species Mouse
Immunogen A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1280, 12824, 25412, 395069, 407142
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.