Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen II Monoclonal Antibody (6B3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA513026
Description
MA5-13026 targets Collagen II in IF and IHC (P) applications and shows reactivity with Amphibian, Chicken, Human, and mouse samples. The MA5-13026 immunogen is purified type II collagen; obtained from the adult chicken sternum by limited pepsinization.
COL2A1 is the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia.
Specifications
Collagen II | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02458, P02460, P28481 | |
Col2a1 | |
Purified type II collagen; obtained from the adult chicken sternum by limited pepsinization | |
500 μL | |
Primary | |
Amphibian, Chicken, Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
6B3 | |
Unconjugated | |
Col2a1 | |
alpha 1 type II procollagen; alpha 1 type IIA collagen; alpha 1 type IIA collagen precursor; alpha collagen type II; alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CB11; CG2A1A; CGA; Chondrocalcin; COL11A3; Col2; Col2a; col2a1; Col2a-1; col2a1.L; col2a1a; col2a1-a; col2a1-b; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen type II alpha 1 chain; collagen type IIA; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); collagen, type II, alpha 1 L homeolog; COLLII; cyanogen bromide; Del1; disproportionate micromelia; Dmm; Lpk; M100856; MGC131516; pro alpha1 type II collagen; pro-alpha 1(II); pro-alpha-1 type II collagen; procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; Rgsc856; SEDC; STL1; type II collagen; type II collagen cyanogen bromide fragment CB10; type II collogen cyanogen bromide fragment CB8; unnamed protein product; X-col II; xcol2a1; XELAEV_18013523mg | |
Mouse | |
Protein G | |
RUO | |
1280, 12824, 395069 | |
4° C | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction