Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Collagen II Native Protein
SDP

Catalog No. NBP197243 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197243 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197243 Supplier Novus Biologicals™ Supplier No. NBP197243
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native COL2A1. Source: Bovine Nasal Cartilage (Accession #: P02459) The Collagen II Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Specifications

Accession Number P02459
For Use With (Application) ELISA, Immunohistochemistry, SDS-PAGE, Western Blot
Formulation 0.5 M Acetic Acid
Gene ID (Entrez) 1280
Name Collagen II protein
Quantity 0.5 mg
Storage Requirements 4°C. Do not freeze.
Cross Reactivity Bovine
Purity or Quality Grade Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.