Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen III alpha 1/COL3A1 Antibody, DyLight 405, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP126547V
Description
Collagen III alpha 1/COL3A1 Polyclonal specifically detects Collagen III alpha 1/COL3A1 in Human samples. It is validated for ELISA.Specifications
Collagen III alpha 1/COL3A1 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
alpha1 (III) collagen, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534 | |
Goat | |
139 kDa | |
0.1 mL | |
Cancer, Cell Biology, Extracellular Matrix | |
Reacts with conformational determinants on human type III collagen as demonstrated by ELISA. May react with type III collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, IV, V and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin). | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
COL3A1 | |
This Collagen III alpha 1/COL3A1 Antibody was developed by hyperimmunizing goats with human type III collagen. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction