Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen III alpha 1/COL3A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215946
Description
Collagen III alpha 1/COL3A1 Polyclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
Collagen III alpha 1/COL3A1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1% BSA, and 20% Glycerol with 0.025% Proclin 300 | |
COL3A1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human Collagen III. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Pig | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
0.31mg/mL | |
Western Blot 1:500 - 1:3000, Immunohistochemistry 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000, Immunohistochemistry-Frozen | |
alpha1 (III) collagen, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534 | |
Rabbit | |
139 kDa | |
0.1 mL | |
Cancer, Cell Biology, Extracellular Matrix | |
1281 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction