Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen III alpha 1/COL3A1 Rabbit anti-Human, Mouse, Rat, Clone: 6Y9C6, Novus Biologicals™
SDP

Catalog No. NB165714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165714 100 μg
NB165715 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165714 Supplier Novus Biologicals Supplier No. NBP315309100UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Collagen III alpha 1/COL3A1 Monoclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation

Specifications

Antigen Collagen III alpha 1/COL3A1
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 6Y9C6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias alpha1 (III) collagen, alpha-1 type III collagen, COL3A1, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, Collagen-III, collagenIII, EDS4A, EDSVASC, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534, PMGEDSV
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-400 of human Collagen III alpha 1/COL3A1 (P02461).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1281
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.