Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen III (COL3A1) Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
SDP

Catalog No. 502543788
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3788 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3788 Supplier OriGene Supplier No. CF812161
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen III (COL3A1) Monoclonal antibody specifically detects Collagen III (COL3A1) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Collagen III (COL3A1)
Applications Western Blot
Classification Monoclonal
Clone OTI2G3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Dnajb4
Gene Accession No. Q9D832, Q9UDY4
Gene Alias alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-337 of human DNAJB4 (NP_008965) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11080, 295549, 67035
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.