Learn More
Invitrogen™ Collagen III Polyclonal Antibody

Description
PA1-28870 has been successfully used in ELISA, immunohistochemistry (frozen tissue), immunoprecipitation, and Western blot applications. NOTE: The preparation of a specific antibody to Collagen III's uninterrupted 'Glycine-X-Y' triplet repeat requires a non-denatured epitope format. Consequently, it is recommended to use this antibody in a Western blot using non-denaturing or 'native' conditions. Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | Collagen III |
| Applications | Dot Blot, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | 1.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 20mM potassium phosphate with 150mM NaCl and 0.01% sodium azide; pH 7.2 |
| Gene | Col3a1 |
| Gene Accession No. | P02461, P04258, P08121 |
| Gene Alias | alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown |
| Gene Symbols | Col3a1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.