Learn More
Invitrogen™ Collagen III Polyclonal Antibody

Description
Immunogen sequence: RGERGSEGSP GHPGQPGPPG PPGAPGPCCG GVGAAAIAGI GGEKAGGFAP YYGDEPMDFK INTDEIMTSL KSVNGQIESL ISPDGSRKNP ARNCRDLKFC HPE Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 93%.
Specifications
Specifications
| Antigen | Collagen III |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Col3a1 |
| Gene Accession No. | P02461 |
| Gene Alias | alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown |
| Gene Symbols | Col3a1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.