Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Collagen III Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA185323
Description
Rabbit anti Rat Collagen III antibody recognizes rat collagen type III, a fibrillar collagen found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen.
Collagen III is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen. Mutations in th gene are associated with Ehlers Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications
Collagen III | |
Polyclonal | |
PBS with 1% antimycotic, <0.1% sodium azide | |
P08121, P13941 | |
Col3a1 | |
purified collagen III from rat skin. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Immunocytochemistry | |
Unconjugated | |
Col3a1 | |
alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12825, 84032 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction