Learn More
Collagen III Polyclonal Antibody, Invitrogen™
Description
Reconstitute with 0.5 mL of distilled water. For long-term storage the addition of 0.09% sodium azide is recommended. Prior to reconstitution, store at 4oC. After reconstitution, store undiluted at -20oC, avoiding freeze/thaw cycles. This antibody has been cross-absorbed with immobilized human collagen types I, IV and V and with human blood plasma proteins. Rabbit anti Human collagen III antibody recognizes human collagen type III.
Specifications
Specifications
| Antigen | Collagen III |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M sodium borate, 0.01M sodium phosphate with 0.1M NaCl, 1% dextran, 1% mannitol and no preservative |
| Gene | Col3a1 |
| Gene Accession No. | P02461 |
| Gene Alias | alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown |
| Gene Symbols | Col3a1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.