Learn More
Invitrogen™ Collagen III Recombinant Rabbit Monoclonal Antibody (6Y9C6)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542628
Description
Positive test controls include: HepG2, RD, SH-SY5Y, Mouse brain, Mouse uterus, Rat lung, Rat uterus. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: NYSPQYDSYD VKSGVAVGGL AGYPGPAGPP GPPGPPGTSG HPGSPGSPGY QGPPGEPGQA GPSGPPGPPG AIGPSGPAGK DGESGRPGRP GERGLPGPPG IKGPAGIPGF PGMKGHRGFD GRNGEKGETG APGLKGENGL PGENGAPGPM GPRGAPGERG RPGLPGAAGA RGNDGARGSD GQPGPPGPPG TAGFPGSPGA KGEVGPAGSP GSNGAPGQRG EPGPQGHAGA QGPPGPPGIN GSPGGKGEMG P.
Collagen III is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen. Mutations in th gene are associated with Ehlers Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications
Collagen III | |
Recombinant Monoclonal | |
0.95 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P02461, P08121, P13941 | |
Col3a1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 150-400 of human Collagen III alpha 1/COL3A1 (P02461). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
6Y9C6 | |
Unconjugated | |
Col3a1 | |
alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1281, 12825, 84032 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.