Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen IV alpha 1 Antibody (577227), Alexa Fluor 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB63082AF700
Description
Collagen IV alpha 1 Monoclonal specifically detects Collagen IV alpha 1 in Human samples. It is validated for Immunocytochemistry.Specifications
Collagen IV alpha 1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
COL4A1 | |
Partial recombinant human Collagen IV alpha 1 protein (amino acids 1441-1669) [UniProt P02462]. | |
0.1 mL | |
Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
Human | |
IgG1 |
Immunocytochemistry | |
577227 | |
Immunocytochemistry | |
alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction