Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen IV Antibody (COL4/4241R), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308277FR
Description
Collagen IV Monoclonal antibody specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Collagen IV | |
Monoclonal | |
DyLight 680 | |
BSVD;BSVD1;COL4A1s;Collagen alpha-1(IV) chain;PADMAL;RATOR | |
Purified human placental extract | |
100 μg | |
Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
1282 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
COL4/4241R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction