Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Mouse anti-Human, Clone: PHM-12, Abnova™

Catalog No. 89933984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-984 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-984 Supplier Abnova Corporation Supplier No. MAB12745

Mouse monoclonal antibody raised against human Collagen IV.

Specifications

Antigen Collagen IV
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PHM-12
Conjugate Unconjugated
Formulation In tissue culture supernatant (0.09% sodium azide).
Host Species Mouse
Immunogen Human Collagen IV
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.