Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Mouse anti-Human, Clone: rCOL4/4742, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Collagen IV
Clone rCOL4/4742
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122666
SDP
View Documents
Novus Biologicals
NBP30759620UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Collagen IV
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers
10 mM PBS with 0.05% BSA
Purified human placental extract
Primary
Store at 4°C.
rCOL4/4742
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
1282
IgG2a κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.