Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Collagen IV Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586939
Description
Purity is > 95% by SDS-PAGE.
Collagen IV is a type of collagen protein that is a major component of the basement membrane in various tissues, including the kidneys, lungs, and skin. It forms a network-like structure that provides support and stability to the surrounding tissue. Collagen IV is composed of three alpha chains, which are encoded by the COL4A1, COL4A2, and COL4A3 genes. Mutations in these genes can lead to various genetic disorders, such as Alport syndrome and Goodpasture syndrome, which affect the kidneys and lungs, respectively.
Specifications
Collagen IV | |
Polyclonal | |
Unconjugated | |
COL4A2 | |
[a]3(IV); [a]4(IV); alpha 1 type IV collagen; alpha type IV collagen basement membrane; alpha1(IV) collagen; alpha-2 collagen type IV; alpha-3 type IV collagen; alpha3(IV); Arresten; Bru; BSVD; CA44; Canstatin; Col4a1; Col4a-1; COL4A1 NC1 domain; Col4a2; Col4a-2; COL4A3; col4a4; Collagen 4; Collagen alpha-1(IV) chain; collagen alpha-2(IV) chain; Collagen alpha-3(IV) chain; collagen alpha-4(IV) chain; collagen IV, alpha-1 polypeptide; collagen IV, alpha-3 polypeptide; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-1 chain; collagen of basement membrane, alpha-4 chain; collagen type IV alpha 1 chain; collagen type IV alpha 2; collagen type IV alpha 2 chain; collagen type IV alpha 3 chain; collagen type IV alpha 4 chain; collagen type IV alpha3 chain; collagen, type IV, alpha 1; collagen, type IV, alpha 2; collagen, type IV, alpha 3; collagen, type IV, alpha 3 (Goodpasture antigen); collagen, type IV, alpha 4; Del(8)44H; Del(8)Bru44H; E130010M05Rik; Goodpasture antigen; ICH; LOW QUALITY PROTEIN: collagen alpha-1(IV) chain; LOW QUALITY PROTEIN: collagen alpha-3(IV) chain; LOW QUALITY PROTEIN: collagen alpha-4(IV) chain; POREN2; procollagen alpha1(IV); procollagen, type IV, alpha 1; procollagen, type IV, alpha 2; procollagen, type IV, alpha 3; procollagen, type IV, alpha 4; RATOR; Raw; retinal anterior wiring; Svc; tumstatin; Type IV Collagen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1282, 1284, 1285, 1286, 1287, 1288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P02462, P08572, P29400, P53420, Q01955, Q14031 | |
COL4A1, COL4A2, Col4a3, COL4A4, COL4A5, COL4A6 | |
Synthetic peptide, corresponding to Human COL4A1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction