Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen IV Rabbit anti-Human, Clone: COL4/4241R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30775020UG
Description
Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Collagen IV | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
COL4/4241R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Purified human placental extract | |
20 μg | |
Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
1282 | |
Store at 4°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction