Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type II Polyclonal Antibody, Rockland™
SDP

Catalog No. 501943827
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-194-3827 500 μg
50-194-3826 100 μg
50-105-7808 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-194-3827 Supplier Rockland Immunochemicals Supplier No. 6004011040.5
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to opening. This product is stable at 4°C as an undiluted liquid. Dilute only prior to immediate use. For extended storage, mix with an equal volume of glycerol, aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. ypically negligible cross reactivity against other types of collagens was detected by ELISA against purified standards. Some class specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type II collagens and has negligible cross-reactivity with Type I, III, IV, V or VI collagens. Non-specific cross reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible.

COL2A1 is the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia.

Specifications

Antigen Collagen Type II
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Col2a1
Gene Accession No. P02458, P02459, P05539, P28481
Gene Alias alpha 1 type II procollagen; alpha 1 type IIA collagen; Alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); BOS_5148; cartilage collagen; CB11; CG2A1A; CGA; Chondrocalcin; COL11A3; Col2; Col2a; COL2A1; Col2a-1; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen type II alpha 1 chain; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); COLLII; cyanogen bromide; Del1; disproportionate micromelia; Dmm; Lpk; M100856; MGC131516; pro-alpha 1(II); procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; Rgsc856; SEDC; STL1
Gene Symbols Col2a1
Host Species Rabbit
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100196905, 1280, 12824, 25412, 407142
Target Species Bovine, Human, Mouse, Ovine, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.