Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen Type XXIII Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Collagen Type XXIII |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9375
![]() ![]() |
Proteintech Group Inc
143371AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
COL23A1 is a member of the transmembrane collagens, a subfamily of the nonfibrillar collagens that contain a single pass hydrophobic transmembrane domain (Banyard et al., 2003 ).Specifications
Collagen Type XXIII | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q810Y4, Q86Y22, Q8K4G2 | |
237759, 353303, 91522 | |
Collagen Type XXIII Fusion Protein Ag5640 | |
Primary | |
-20°C | |
COL23A1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COL23A1, Collagen alpha 1(XXIII) chain, Collagen Type XXIII, collagen, type XXIII, alpha 1 | |
Col23a1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title