Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen V Alpha-1 Chain (NC2 domain) Monoclonal Antibody (13F6), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 90010100UG

This item is not returnable.
View return policy
Description
Collagen V Alpha-1 Chain (NC2 domain) Monoclonal antibody specifically detects Collagen V Alpha-1 Chain (NC2 domain) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Collagen V Alpha-1 Chain (NC2 domain) | |
Monoclonal | |
0.5-1.0 mg/mL | |
0.1M tris with 0.1M glycine, 2% sucrose and no preservative | |
P20908 | |
Mouse | |
Protein A | |
RUO | |
1289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
13F6 | |
Unconjugated | |
COL5A1 | |
AI413331; alpha 1 (V) collagen; alpha1(V) collagen; cha1V; COL5A1; collagen alpha 1 (V); collagen alpha-1(V) chain; collagen type V alpha 1; collagen type V alpha 1 chain; collagen, type V, alpha 1; EDSC; pro-alpha1(V) collagen; procollagen type V alpha 1; procollagen, type V, alpha 1; type V collagen; type V collagen, alpha 1 chain, NH2-terminal domain | |
Recombinant human collagen V alpha-1 chain N fragment. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction