Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collagen V Monoclonal Antibody (MC5-HA)
GREENER_CHOICE

Catalog No. PIMA126150
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126150 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126150 Supplier Invitrogen™ Supplier No. MA126150
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-26150 detects Collagen V from human samples.

COL5A1 encodes an alpha chain for one of the lowabundance fibrillar collagens. Fibrillar collagen molecules aretrimers that can be composed of one or more types of alpha chains.Type V collagen is found in tissues containing type I collagen andappears to regulate the assembly of heterotypic fibers composed ofboth type I and type V collagen. COL5A1 product is closelyrelated to type XI collagen and it is possible that the collagenchains of types V and XI constitute a single collagen type withtissue-specific chain combinations. Mutations in this gene areassociated with Ehlers-Danlos syndrome, types I and II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen V
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone MC5-HA
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS, 10mM sodium borate with 1% dextran, 1% mannitol and no preservative; pH 8
Gene COL5A1
Gene Accession No. P20908
Gene Alias AB collagen; AI413331; alpha-1 type V collagen; alpha1(V) collagen; COL5; Col5a1; Collagen 5; collagen alpha 1 (V); collagen alpha-1(V) chain; collagen type 5 alpha 1; collagen type V alpha 1; collagen type V alpha 1 chain; collagen, type V, alpha 1; EDSC; pro-alpha1(V) collagen; procollagen alpha 1(V); procollagen type V alpha 1; procollagen, type V, alpha 1; RP11-263F14.1; Type V Collagen
Gene Symbols COL5A1
Host Species Mouse
Immunogen Full length native protein (purified) (Human).
Purification Method IgM
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1289
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.