Learn More
Description
Specifications
Specifications
| Formulation | 0.5 M Acetic Acid |
| Gene ID (Entrez) | 1291 |
| Name | Collagen VI protein |
| Quantity | 0.5 mg |
| Source | Bovine Serum |
| Storage Requirements | 4°C. Do not freeze. |
| Cross Reactivity | Bovine |
| Purity or Quality Grade | Chromatography |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
