Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen VI alpha 3 Antibody (118), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290328MFV610
Description
Collagen VI alpha 3 Monoclonal antibody specifically detects Collagen VI alpha 3 in Human samples. It is validated for ELISASpecifications
Collagen VI alpha 3 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
1293 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
118 | |
ELISA | |
Collagen Alpha-3(VI) Chain, Collagen VI, Alpha-3 Polypeptide, collagen, type VI, alpha 3 | |
Rabbit immunized with purified, recombinant Human Collagen VI alpha 3 (Uniprot#: P12111-1, 3101T-3177T) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction