Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collagen VI Recombinant Rabbit Monoclonal Antibody (7J1P4)
GREENER_CHOICE

Catalog No. PIMA538207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538207 Supplier Invitrogen™ Supplier No. MA538207
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse lung, Rat heart Immunogen sequence: DLFFVLDTSE SVALRLKPYG ALVDKVKSFT KRFIDNLRDR YYRCDRNLVW NAGALHYSDE VEIIQGLTRM PGGRDALKSS VDAVKYFGKG TYTDCAIKKG LEQLLVGGSH LKENKYLIVV TDGHPLEGYK EPCGGLEDAV NEAKHLGVKV FSVAITPDHL EPRLSIIATD HTYRRNFTAA DWGQSRDAEE AISQTIDTI.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen VI
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7J1P4
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide, pH 7.3
Gene COL6A1
Gene Accession No. P12109, Q04857
Gene Alias AI747156; alpha 1 (VI) chain (61 AA); BTHLM1; Col6a1; Col6a-1; Col6a2; Col6a-2; Collagen alpha-1(VI) chain; collagen alpha-2(VI) chain; collagen type VI alpha 1; collagen type VI alpha 2; collagen VI, alpha-1 polypeptide; collagen VI, alpha-2 polypeptide; collagen, type VI, alpha 1; collagen, type VI, alpha 2; human mRNA for collagen VI alpha-2 C-terminal globular domain; OPLL; PP3610; procollagen, type VI, alpha 1; procollagen, type VI, alpha 2; RGD1565398; UCHMD1; UCMD1
Gene Symbols COL6A1
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12833, 1291, 294337
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.