Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collagen VI Recombinant Rabbit Monoclonal Antibody (SD83-03)
GREENER_CHOICE

Catalog No. PIMA532412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532412 Supplier Invitrogen™ Supplier No. MA532412
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen VI
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SD83-03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene COL6A1
Gene Accession No. P12109, Q04857
Gene Alias AI747156; alpha 1 (VI) chain (61 AA); BTHLM1; Col6a1; Col6a-1; Col6a2; Col6a-2; Collagen alpha-1(VI) chain; collagen alpha-2(VI) chain; collagen type VI alpha 1; collagen type VI alpha 2; collagen VI, alpha-1 polypeptide; collagen VI, alpha-2 polypeptide; collagen, type VI, alpha 1; collagen, type VI, alpha 2; human mRNA for collagen VI alpha-2 C-terminal globular domain; OPLL; PP3610; procollagen, type VI, alpha 1; procollagen, type VI, alpha 2; RGD1565398; UCHMD1; UCMD1
Gene Symbols COL6A1
Host Species Rabbit
Immunogen Recombinant protein within Human Collagen VI aa 17-255.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12833, 1291, 294337
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.