Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen VII Antibody (LH7.2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308278H
Description
Collagen VII Monoclonal antibody specifically detects Collagen VII in Human, Bovine, Equine, Guinea Pig, Goat, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Collagen VII | |
Monoclonal | |
HRP | |
Collagen 7, collagen alpha-1(VII) chain, collagen VII, alpha-1 polypeptide, collagen, type VII, alpha 1, Collagen-7, EBD1, EBDCT, EBR1, epidermolysis bullosa, dystrophic, dominant and recessive, LC collagen, long chain collagen, Long-chain collagen | |
Recombinant full-length human Collagen VII protein (Uniprot: Q02388) | |
100 μg | |
Cell Biology, Cellular Markers, Extracellular Matrix | |
1294 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
LH7.2 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Equine, Guinea Pig, Goat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction