Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen VII Mouse anti-Human, Bovine, Equine, Guinea Pig, Goat, Sheep, Clone: LH7.2, Novus Biologicals™
SDP

Catalog No. NB407833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB407833 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB407833 Supplier Novus Biologicals Supplier No. NBP307751100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen VII Monoclonal specifically detects Collagen VII in Human, Bovine, Equine, Guinea Pig, Goat, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Collagen VII
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone LH7.2
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin
Formulation 10 mM PBS with 0.05% BSA
Gene Alias Collagen 7, collagen alpha-1(VII) chain, collagen VII, alpha-1 polypeptide, collagen, type VII, alpha 1, Collagen-7, EBD1, EBDCT, EBR1, epidermolysis bullosa, dystrophic, dominant and recessive, LC collagen, long chain collagen, Long-chain collagen
Host Species Mouse
Immunogen Recombinant full-length human Collagen VII protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1294
Target Species Human, Bovine, Equine, Guinea Pig, Goat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.