Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen X alpha 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30375720UL
Description
Collagen X alpha 1 Polyclonal specifically detects Collagen X alpha 1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Collagen X alpha 1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
1300 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry | |
Unconjugated | |
COL10A1, Collagen Alpha-1(X) Chain, Collagen X, Alpha-1 Polypeptide, Schmid Metaphyseal Chondrodysplasia | |
Recombinant fusion protein containing a sequence corresponding to amino acids 521-680 of human Collagen X alpha 1 (NP_000484.2). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction