Learn More
Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$186.65
Specifications
Antigen | Collagen X |
---|---|
Clone | X53 |
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
41-977-180
![]() |
Invitrogen™
41977180 |
25 μg |
$196.00
$186.65 / Each of 1 Save $9.35 5% Off |
|
|||||
Description
The MAb X53 recognizes human, mouse, porcine, guinea pig, and rat Collagen Type X. X53 does not cross-react with collagen types I, II, II, IV, or VI. Collagen Type X is a homotrimeric short chain collagen. Collagen Type X has limited expression in normal adult tissues, primarily being expressed during bone growth and development. Expression is localized to the peri- and extracellular matrix of hypertrophic chondrocytes during endochondral ossification and also plays a role in cartilage mineralization. Collagen Type X is elevated in many tumor types with expression being localized in the vasculature. Mutations to the gene COL10A1, which encodes human Collagen Type X, is associated with Schmid type metaphyseal chondrodysplasia.
Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.Specifications
Collagen X | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Col10; COL10A1; Col10a-1; Collagen alpha-1(X) chain; collagen type X alpha 1 chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1 | |
COL10A1 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
COL10A1 |
X53 | |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry | |
eFluor 570 | |
Mouse | |
Canine, Guinea Pig, Human, Mouse, Pig, Rat | |
Q03692, Q05306 | |
100856711, 106026924, 12813, 1300, 25681, 448809 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.