Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen XI alpha 2 Antibody (6410), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243809
Description
Ensure accurate, reproducible results in Western Blot
Collagen XI alpha 2 Monoclonal antibody specifically detects Collagen XI alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Collagen XI alpha 2 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
COL11A2 | |
Recombinant protein encompassing a sequence within the N-terminus region of human COL11A2. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
6410 | |
Western Blot 1:500-1:3000 | |
collagen alpha-2(XI) chain, collagen, type XI, alpha 2, DFNA13, DFNB53, HKE5, PARP, pro-a2 chain of collagen type XI, STL3 | |
Mouse | |
Protein G purified | |
RUO | |
1302 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction