Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen XI alpha 2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Collagen XI alpha 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159276
SDP
View Documents
Novus Biologicals
NBP159276
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Collagen XI alpha 2 Polyclonal specifically detects Collagen XI alpha 2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Collagen XI alpha 2
Polyclonal
Rabbit
P13942
1302
Synthetic peptides corresponding to COL11A2(collagen, type XI, alpha 2) The peptide sequence was selected from the N terminal of COL11A2. Peptide sequence TADRFQAEEYGEGGTDPPEGPYDYTYGYGDDYREETELGPALSAETAHSG.
Primary
Western Blot
Unconjugated
RUO
collagen alpha-2(XI) chain, collagen, type XI, alpha 2, DFNA13, DFNB53, HKE5, PARP, pro-a2 chain of collagen type XI, STL3
COL11A2
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.